Fmoc-(S)-3-amino-4-(3-trifluoromethylphenyl)butyric acid

Supplier: Matrix Scientific

Synonyms: (S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(3-(trifluoromethyl)phenyl)butanoic acid

042353-250MG 042353-1G
101855-474EA 412.31 USD
101855-474 101855-476
Fmoc-(S)-3-amino-4-(3-trifluoromethylphenyl)butyric acid
Fmoc-(S)-3-amino-4-(3-trifluoromethylphenyl)butyric acid

Hazard: Irritant

Formula: C₂₆H₂₂F₃NO₄
MW: 469.46 g/mol
MDL Number: MFCD01861068
CAS Number: 270065-78-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR