Fmoc-(S)-3-amino-4-(3-thienyl)butyric acid
Supplier: Matrix Scientific
042352-250MG
042352-1G
101855-470EA
412.31
USD
101855-470
101855-472
Fmoc-(S)-3-amino-4-(3-thienyl)butyric acid
Fmoc-(S)-3-amino-4-(3-thienyl)butyric acid
Hazard: Irritant
Formula:
C₂₃H₂₁NO₄S |
MDL Number:
MFCD01861085 CAS Number: 270263-01-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...