Fmoc-(S)-3-amino-4-(3-fluorophenyl)butyric acid

Supplier: Matrix Scientific

Synonyms: (S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(3-fluorophenyl)butanoic acid

042349-250MG 042349-1G
101855-458EA 275.38 USD
101855-458 101855-460
Fmoc-(S)-3-amino-4-(3-fluorophenyl)butyric acid
Fmoc-(S)-3-amino-4-(3-fluorophenyl)butyric acid

Hazard: Irritant

Formula: C₂₅H₂₂FNO₄
MW: 419.46 g/mol
MDL Number: MFCD02094568
CAS Number: 270596-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR