Fmoc-(S)-3-amino-4-(3-benzothienyl)butyric acid

Supplier: Matrix Scientific

Synonyms: (S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(benzo[b]thiophen-3-yl)butanoic acid

042346-250MG 042346-1G
101855-446EA 349.23 USD
101855-446 101855-448
Fmoc-(S)-3-amino-4-(3-benzothienyl)butyric acid
Fmoc-(S)-3-amino-4-(3-benzothienyl)butyric acid

Hazard: Irritant

Formula: C₂₇H₂₃NO₄S
MW: 457.55 g/mol
MDL Number: MFCD01861042
CAS Number: 270063-46-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR