Fmoc-(S)-3-amino-4-(3,4-difluorophenyl)butyric acid

Supplier: Matrix Scientific

Synonyms: (S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(3,4-difluorophenyl)butanoic acid

042345-250MG 042345-1G
101855-442EA 376.92 USD
101855-442 101855-444
Fmoc-(S)-3-amino-4-(3,4-difluorophenyl)butyric acid
Fmoc-(S)-3-amino-4-(3,4-difluorophenyl)butyric acid

Hazard: Irritant

Formula: C₂₅H₂₁F₂NO₄
MW: 437.45 g/mol
MDL Number: MFCD01861051
CAS Number: 270063-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR