Fmoc-(S)-3-amino-4-(3,4-dichlorophenyl)butyricacid

Supplier: Matrix Scientific

Synonyms: (S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(3,4-dichlorophenyl)butanoic acid

042344-250MG 042344-1G
101855-438EA 349.23 USD
101855-438 101855-440
Fmoc-(S)-3-amino-4-(3,4-dichlorophenyl)butyricacid
Fmoc-(S)-3-amino-4-(3,4-dichlorophenyl)butyricacid

Hazard: Irritant

Formula: C₂₅H₂₁Cl₂NO₄
MW: 470.36 g/mol
MDL Number: MFCD01861048
CAS Number: 270063-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR