Fmoc-(S)-3-amino-4-(2-naphthyl)butyric acid

Supplier: Matrix Scientific

Synonyms: (S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(naphthalen-2-yl)butanoic acid

042341-250MG 042341-1G
101855-426EA 275.38 USD
101855-426 101855-428
Fmoc-(S)-3-amino-4-(2-naphthyl)butyric acid
Fmoc-(S)-3-amino-4-(2-naphthyl)butyric acid

Hazard: Irritant

Formula: C₂₉H₂₅NO₄
MW: 451.53 g/mol
MDL Number: MFCD01861036
CAS Number: 1186216-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR