Fmoc-(S)-3-amino-4-(2-methylphenyl)butyric acid

Supplier: Matrix Scientific

Synonyms: (S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(o-tolyl)butanoic acid

042340-250MG 042340-1G
101855-422EA 275.38 USD
101855-422 101855-424
Fmoc-(S)-3-amino-4-(2-methylphenyl)butyric acid
Fmoc-(S)-3-amino-4-(2-methylphenyl)butyric acid

Hazard: Irritant

Formula: C₂₆H₂₅NO₄
MW: 415.49 g/mol
MDL Number: MFCD01861021
CAS Number: 270062-91-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR