Fmoc-(S)-3-amino-4-(2-furyl)butyric acid

Supplier: Matrix Scientific

042339-250MG 042339-1G
101855-418EA 498.46 USD
101855-418 101855-420
Fmoc-(S)-3-amino-4-(2-furyl)butyric acid
Fmoc-(S)-3-amino-4-(2-furyl)butyric acid

Hazard: Irritant

Formula: C₂₃H₂₁NO₅
MDL Number: MFCD01861091
CAS Number: 270263-07-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR