Fmoc-(S)-3-amino-4-(2-fluorophenyl)butyric acid

Supplier: Matrix Scientific

Synonyms: (S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(2-fluorophenyl)butanoic acid

042338-250MG 042338-1G
101855-414EA 275.38 USD
101855-414 101855-416
Fmoc-(S)-3-amino-4-(2-fluorophenyl)butyric acid
Fmoc-(S)-3-amino-4-(2-fluorophenyl)butyric acid

Hazard: Irritant

Formula: C₂₅H₂₂FNO₄
MW: 419.46 g/mol
MDL Number: MFCD01861009
CAS Number: 270596-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR