Fmoc-(S)-3-amino-4-(2-cyanophenyl)butyric acid

Supplier: Matrix Scientific

042337-250MG 042337-1G
101855-410EA 412.31 USD
101855-410 101855-412
Fmoc-(S)-3-amino-4-(2-cyanophenyl)butyric acid
Fmoc-(S)-3-amino-4-(2-cyanophenyl)butyric acid

Hazard: Irritant

Formula: C₂₆H₂₂N₂O₄
MDL Number: MFCD01861074
CAS Number: 270065-84-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR