Fmoc-(S)-3-amino-4-(2,4-dichlorophenyl)butyricacid

Supplier: Matrix Scientific

042335-250MG 042335-1G
101855-402EA 349.23 USD
101855-402 101855-404
Fmoc-(S)-3-amino-4-(2,4-dichlorophenyl)butyricacid
Fmoc-(S)-3-amino-4-(2,4-dichlorophenyl)butyricacid

Hazard: Irritant

Formula: C₂₅H₂₁Cl₂NO₄
MDL Number: MFCD01861045
CAS Number: 270063-49-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR