Fmoc-(S)-3-amino-4-(1-naphthyl)butyric acid

Supplier: Matrix Scientific

042334-250MG 042334-1G
101855-398EA 275.38 USD
101855-398 101855-400
Fmoc-(S)-3-amino-4-(1-naphthyl)butyric acid
Fmoc-(S)-3-amino-4-(1-naphthyl)butyric acid

Hazard: Irritant

Formula: C₂₉H₂₅NO₄
MDL Number: MFCD01861034
CAS Number: 270063-38-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR