Fmoc-(R)-3-amino-6-phenyl-5-hexenoic acid

Supplier: Matrix Scientific

042331-250MG 042331-1G
101855-386EA 412.31 USD
101855-386 101855-388
Fmoc-(R)-3-amino-6-phenyl-5-hexenoic acid
Fmoc-(R)-3-amino-6-phenyl-5-hexenoic acid

Hazard: Irritant

Formula: C₂₇H₂₅NO₄
MDL Number: MFCD01860907
CAS Number: 332064-75-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR