Fmoc-(R)-3-amino-5-hexynoic acid
Supplier: Matrix Scientific
Synonyms:
(R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)hex-5-ynoic acid
042329-250MG
042329-1G
101855-378EA
461.54
USD
101855-378
101855-380
Fmoc-(R)-3-amino-5-hexynoic acid
Fmoc-(R)-3-amino-5-hexynoic acid
Hazard: Irritant
Formula:
C₂₁H₁₉NO₄ MW: 349.39 g/mol |
MDL Number:
MFCD01860910 CAS Number: 332064-94-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...