Fmoc-(R)-3-amino-5-hexenoic acid

Supplier: Matrix Scientific

Synonyms: (R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)hex-5-enoic acid

042328-250MG 042328-1G
101855-374EA 412.31 USD
101855-374 101855-376
Fmoc-(R)-3-amino-5-hexenoic acid
Fmoc-(R)-3-amino-5-hexenoic acid

Hazard: Irritant

Formula: C₂₁H₂₁NO₄
MW: 351.41 g/mol
MDL Number: MFCD01860991
CAS Number: 269726-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR