Fmoc-(R)-3-amino-4-(4-pyridyl)butyric acid

Supplier: Matrix Scientific

042324-250MG 042324-1G
101855-358EA 510.77 USD
101855-358 101855-360
Fmoc-(R)-3-amino-4-(4-pyridyl)butyric acid
Fmoc-(R)-3-amino-4-(4-pyridyl)butyric acid

Hazard: Irritant

Formula: C₂₄H₂₂N₂O₄
MDL Number: MFCD01860961
CAS Number: 269396-69-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR