Fmoc-(R)-3-amino-4-(4-methylphenyl)butyric acid
Supplier: Matrix Scientific
042322-250MG
042322-1G
101855-350EA
155.38
USD
101855-350
101855-352
Fmoc-(R)-3-amino-4-(4-methylphenyl)butyric acid
Fmoc-(R)-3-amino-4-(4-methylphenyl)butyric acid
Hazard: Irritant
Formula:
C₂₆H₂₅NO₄ |
MDL Number:
MFCD01860929 CAS Number: 269398-86-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...