Fmoc-(R)-3-amino-4-(4-chlorophenyl)butyric acid

Supplier: Matrix Scientific

Synonyms: (R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(4-chlorophenyl)butanoic acid

042318-250MG 042318-1G
101855-334EA 275.38 USD
101855-334 101855-336
Fmoc-(R)-3-amino-4-(4-chlorophenyl)butyric acid
Fmoc-(R)-3-amino-4-(4-chlorophenyl)butyric acid

Hazard: Irritant

Formula: C₂₅H₂₂ClNO₄
MW: 435.91 g/mol
MDL Number: MFCD01860904
CAS Number: 331763-60-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR