Fmoc-(R)-3-amino-4-(4-bromophenyl)butyric acid
Supplier: Matrix Scientific
Synonyms:
(R)-3-((((9H-Ffluoren-9-yl)methoxy)carbonyl)amino)-4-(4-bromophenyl)butanoic acid
042317-1G
042317-5G
101855-330EA
821.54
USD
101855-330
101855-332
Fmoc-(R)-3-amino-4-(4-bromophenyl)butyric acid
Fmoc-(R)-3-amino-4-(4-bromophenyl)butyric acid
Hazard: Irritant
Formula:
C₂₅H₂₂BrNO₄ MW: 480.37 g/mol |
MDL Number:
MFCD01860918 CAS Number: 331763-76-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...