Fmoc-(R)-3-amino-4-(3-trifluoromethylphenyl)butyric acid

Supplier: Matrix Scientific

Synonyms: (R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(3-(trifluoromethyl)phenyl)butanoic acid

042316-250MG 042316-1G
101855-326EA 412.31 USD
101855-326 101855-328
Fmoc-(R)-3-amino-4-(3-trifluoromethylphenyl)butyric acid
Fmoc-(R)-3-amino-4-(3-trifluoromethylphenyl)butyric acid

Hazard: Irritant

Formula: C₂₆H₂₂F₃NO₄
MW: 469.46 g/mol
MDL Number: MFCD01860970
CAS Number: 269726-75-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR