Fmoc-(R)-3-amino-4-(3-thienyl)butyric acid
Supplier: Matrix Scientific
042315-250MG
042315-1G
101855-322EA
412.31
USD
101855-322
101855-324
Fmoc-(R)-3-amino-4-(3-thienyl)butyric acid
Fmoc-(R)-3-amino-4-(3-thienyl)butyric acid
Hazard: Irritant
Formula:
C₂₃H₂₁NO₄S |
MDL Number:
MFCD01860988 CAS Number: 269726-93-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...