Fmoc-(R)-3-amino-4-(3-fluorophenyl)butyric acid

Supplier: Matrix Scientific

Synonyms: (R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(3-fluorophenyl)butanoic acid

042312-250MG 042312-1G
101855-310EA 275.38 USD
101855-310 101855-312
Fmoc-(R)-3-amino-4-(3-fluorophenyl)butyric acid
Fmoc-(R)-3-amino-4-(3-fluorophenyl)butyric acid

Hazard: Irritant

Formula: C₂₅H₂₂FNO₄
MW: 419.46 g/mol
MDL Number: MFCD02094580
CAS Number: 331763-67-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR