Fmoc-(R)-3-amino-4-(3-cyanophenyl)butyric acid

Supplier: Matrix Scientific

Synonyms: (R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(3-cyanophenyl)butanoic acid

042311-250MG 042311-1G
101855-306EA 412.31 USD
101855-306 101855-308
Fmoc-(R)-3-amino-4-(3-cyanophenyl)butyric acid
Fmoc-(R)-3-amino-4-(3-cyanophenyl)butyric acid

Hazard: Irritant

Formula: C₂₆H₂₂N₂O₄
MW: 426.48 g/mol
MDL Number: MFCD01860979
CAS Number: 269726-84-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR