Fmoc-(R)-3-amino-4-(3-benzothienyl)butyric acid

Supplier: Matrix Scientific

042309-250MG 042309-1G
101855-298EA 349.23 USD
101855-298 101855-300
Fmoc-(R)-3-amino-4-(3-benzothienyl)butyric acid
Fmoc-(R)-3-amino-4-(3-benzothienyl)butyric acid

Hazard: Irritant

Formula: C₂₇H₂₃NO₄S
MDL Number: MFCD01860944
CAS Number: 269396-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR