Fmoc-(R)-3-amino-4-(3,4-difluorophenyl)butyricacid
Supplier: Matrix Scientific
Synonyms:
(R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(3,4-difluorophenyl)butanoic acid
042308-250MG
042308-1G
101855-294EA
412.31
USD
101855-294
101855-296
Fmoc-(R)-3-amino-4-(3,4-difluorophenyl)butyricacid
Fmoc-(R)-3-amino-4-(3,4-difluorophenyl)butyricacid
Hazard: Irritant
Formula:
C₂₅H₂₁F₂NO₄ MW: 437.45 g/mol |
MDL Number:
MFCD01860953 CAS Number: 269396-60-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...