Fmoc-(R)-3-amino-4-(3,4-dichlorophenyl)butyricacid

Supplier: Matrix Scientific

Synonyms: Fmoc-D-β-HoPhe(3,4-DiCl)-OH, (R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(3,4-dichlorophenyl)butanoic acid

042307-250MG 042307-1G
101855-290EA 349.23 USD
101855-290 101855-292
Fmoc-(R)-3-amino-4-(3,4-dichlorophenyl)butyricacid
Fmoc-(R)-3-amino-4-(3,4-dichlorophenyl)butyricacid

Hazard: Irritant

Formula: C₂₅H₂₁Cl₂NO₄
MW: 470.36 g/mol
MDL Number: MFCD01860950
CAS Number: 269396-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR