Fmoc-(R)-3-amino-4-(2-trifluoromethylphenyl)butyric acid

Supplier: Matrix Scientific

042306-250MG 042306-1G
101855-286EA 412.31 USD
101855-286 101855-288
Fmoc-(R)-3-amino-4-(2-trifluoromethylphenyl)butyric acid
Fmoc-(R)-3-amino-4-(2-trifluoromethylphenyl)butyric acid

Hazard: Irritant

Formula: C₂₆H₂₂F₃NO₄
MDL Number: MFCD01860967
CAS Number: 269726-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR