Fmoc-(R)-3-amino-4-(2-fluorophenyl)butyric acid

Supplier: Matrix Scientific

Synonyms: (R)-3-(Fmoc-amino)-4-(2-fluorophenyl)butanoic acid

042301-250MG 042301-1G
101855-266EA 275.38 USD
101855-266 101855-268
Fmoc-(R)-3-amino-4-(2-fluorophenyl)butyric acid
Fmoc-(R)-3-amino-4-(2-fluorophenyl)butyric acid

Hazard: Irritant

Formula: C₂₅H₂₂FNO₄
MW: 419.46 g/mol
MDL Number: MFCD01860911
CAS Number: 331763-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR