Fmoc-(R)-3-amino-4-(2,4-dichlorophenyl)butyric acid

Supplier: Matrix Scientific

Synonyms: Fmoc-D-β-HoPhe(2,4-DiCl)-OH, (R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(2,4-dichlorophenyl)butanoic acid

042298-250MG 042298-1G
101855-254EA 349.23 USD
101855-254 101855-256
Fmoc-(R)-3-amino-4-(2,4-dichlorophenyl)butyric acid
Fmoc-(R)-3-amino-4-(2,4-dichlorophenyl)butyric acid

Hazard: Irritant

Formula: C₂₅H₂₁Cl₂NO₄
MW: 470.36 g/mol
MDL Number: MFCD01860947
CAS Number: 269396-54-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR