Fmoc-(R)-3-amino-4-(1-naphthyl)butyric acid

Supplier: Matrix Scientific

042297-250MG 042297-1G
101855-250EA 275.38 USD
101855-250 101855-252
Fmoc-(R)-3-amino-4-(1-naphthyl)butyric acid
Fmoc-(R)-3-amino-4-(1-naphthyl)butyric acid

Hazard: Irritant

Formula: C₂₉H₂₅NO₄
MDL Number: MFCD01860936
CAS Number: 269398-89-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR