Fmoc-(R)-1,2,3,4-tetrahydro-isoquinoline-3-aceticacid

Supplier: Matrix Scientific

Synonyms: (R)-2-(2-(((9H-Fluoren-9-yl)methoxy)carbonyl)-1,2,3,4-tetrahydroisoquinolin-3-yl)acetic acid

042296-250MG 042296-1G
101855-246EA 318.46 USD
101855-246 101855-248
Fmoc-(R)-1,2,3,4-tetrahydro-isoquinoline-3-aceticacid
Fmoc-(R)-1,2,3,4-tetrahydro-isoquinoline-3-aceticacid

Hazard: Irritant

Formula: C₂₆H₂₃NO₄
MW: 413.48 g/mol
MDL Number: MFCD01860934
CAS Number: 332064-67-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR