Boc-O-benzyl-L-β-homotyrosine
Supplier: Matrix Scientific
Synonyms:
Boc-β-HoTyr(OBzl)-OH, (S)-4-(4-(Benzyloxy)phenyl)-3-((tert-butoxycarbonyl)amino)butanoic acid
042295-250MG
042295-1G
101855-242EA
349.23
USD
101855-242
101855-244
Boc-O-benzyl-L-β-homotyrosine
Boc-O-benzyl-L-beta-homotyrosine
Hazard: Irritant
Formula:
C₂₂H₂₇NO₅ MW: 385.46 g/mol |
MDL Number:
MFCD01862863 CAS Number: 126825-16-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...