Boc-O-benzyl-L-β-homothreonine

Supplier: Matrix Scientific

Synonyms: (3R,4R)-4-(Benzyloxy)-3-((tert-butoxycarbonyl)amino)pentanoic acid

042294-250MG 042294-1G
101855-238EA 287.69 USD
101855-238 101855-240
Boc-O-benzyl-L-β-homothreonine
Boc-O-benzyl-L-beta-homothreonine

Hazard: Irritant

Formula: C₁₇H₂₅NO₅
MW: 323.39 g/mol
MDL Number: MFCD01862935
CAS Number: 254101-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR