Boc-O-benzyl-L-β-homoserine

Supplier: Matrix Scientific

Synonyms: Boc-β-HomoSer(Bzl)-OH, Boc-β-HoSer(Bzl)-OH, Boc-β-Hse(Bzl)-OH

042293-250MG 042293-1G
101855-234EA 349.23 USD
101855-234 101855-236
Boc-O-benzyl-L-β-homoserine
Boc-O-benzyl-L-beta-homoserine

Hazard: Irritant

Formula: C₁₆H₂₃NO₅
MW: 309.37 g/mol
MDL Number: MFCD01862866
CAS Number: 218943-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR