Boc-O-benzyl-L-β-homohydroxyproline

Supplier: Matrix Scientific

042292-100MG 042292-500MG
101855-230EA 275.38 USD
101855-230 101855-232
Boc-O-benzyl-L-β-homohydroxyproline
Boc-O-benzyl-L-beta-homohydroxyproline

Hazard: Irritant

Formula: C₁₈H₂₅NO₅
MW: 335.4 g/mol
MDL Number: MFCD12547722
CAS Number: 136271-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR