Boc-Nw-tosyl-L-β-homoarginine

Supplier: Matrix Scientific

Synonyms: Boc-β-HomoArg(Tos)-OH, Boc-β-HoArg(Tos)-OH

042291-250MG 042291-1G
101855-226EA 412.31 USD
101855-226 101855-228
Boc-Nw-tosyl-L-β-homoarginine
Boc-Nw-tosyl-L-beta-homoarginine

Hazard: Irritant

Formula: C₁₉H₃₀N₄O₆S
MW: 442.54 g/mol
MDL Number: MFCD03427576
CAS Number: 136271-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR