Boc-L-Nw-benzyloxycarbonyl-β-homolysine

Supplier: Matrix Scientific

Synonyms: (S)-7-(((Benzyloxy)carbonyl)amino)-3-((tert-butoxycarbonyl)amino)heptanoic acid, Boc-β-HoLys(Cbz)-OH

042290-250MG 042290-1G
101855-222EA 275.38 USD
101855-222 101855-224
Boc-L-Nw-benzyloxycarbonyl-β-homolysine
Boc-L-Nw-benzyloxycarbonyl-beta-homolysine

Hazard: Irritant

Formula: C₂₀H₃₀N₂O₆
MW: 394.47 g/mol
MDL Number: MFCD01862937
CAS Number: 346694-77-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR