(S)-2-(1-(tert-Butoxycarbonyl)pyrrolidin-2-yl)acetic acid

Supplier: Matrix Scientific

Synonyms: (S)-2-(1-BOC-2-pyrrolidinyl)acetic acid, Boc-β-HomoPro-OH

042287-250MG 042287-1G
101855-210EA 243.08 USD
101855-210 101855-212
(S)-2-(1-(tert-Butoxycarbonyl)pyrrolidin-2-yl)acetic acid
(S)-2-(1-(tert-Butoxycarbonyl)pyrrolidin-2-yl)acetic acid

Hazard: Irritant

Formula: C₁₁H₁₉NO₄
MW: 229.3 g/mol
MDL Number: MFCD01862939
CAS Number: 56502-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR