Boc-L-β-homomethionine

Supplier: Matrix Scientific

Synonyms: (R)-3-((tert-Butoxycarbonyl)amino)-5-(methylthio)pentanoic acid, Boc-β-HoMet-OH

042285-250MG 042285-1G
101855-202EA 238.46 USD
101855-202 101855-204
Boc-L-β-homomethionine
Boc-L-beta-homomethionine

Hazard: Irritant

Formula: C₁₁H₂₁NO₄S
MW: 263.36 g/mol
MDL Number: MFCD01862845
CAS Number: 244251-20-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR