Boc-L-β-homoleucine

Supplier: Matrix Scientific

Synonyms: Boc-β-HoLeu-OH, (S)-3-((tert-Butoxycarbonyl)amino)-5-methylhexanoic acid

042284-250MG 042284-1G
101855-198EA 238.46 USD
101855-198 101855-200
Boc-L-β-homoleucine
Boc-L-beta-homoleucine

Hazard: Irritant

Formula: C₁₂H₂₃NO₄
MW: 245.32 g/mol
MDL Number: MFCD02101665
CAS Number: 132549-43-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR