Boc-L-β-homoisoleucine

Supplier: Matrix Scientific

Synonyms: Boc-β-HomoIle-OH, Boc-β-HoIle-OH

042283-1G 042283-5G
101855-194EA 661.54 USD
101855-194 101855-196
Boc-L-β-homoisoleucine
Boc-L-beta-homoisoleucine

Hazard: Irritant

Formula: C₁₂H₂₃NO₄
MW: 245.32 g/mol
MDL Number: MFCD01076254
CAS Number: 218608-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR