Boc-L-β-homoglutamic acid-d-benzyl ester

Supplier: Matrix Scientific

Synonyms: (S)-6-(Benzyloxy)-3-((tert-butoxycarbonyl)amino)-6-oxohexanoic acid, Boc-β-HomoGlu(OBzl)-OH, Boc-β-HoGlu(OBzl)-OH

042282-250MG 042282-1G
101855-190EA 287.69 USD
101855-190 101855-192
Boc-L-β-homoglutamic acid-d-benzyl ester
Boc-L-beta-homoglutamic acid-d-benzyl ester

Hazard: Irritant

Formula: C₁₈H₂₅NO₆
MW: 351.4 g/mol
MDL Number: MFCD01862936
CAS Number: 218943-30-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR