Boc-L-β-homoaspartic acid-γ-benzyl ester

Supplier: Matrix Scientific

Synonyms: Boc-β-HomoAsp(OBzl)-OH, Boc-β-HoAsp(OBzl)-OH

042281-1G 042281-250MG
101855-186EA 823.08 USD
101855-186 101855-188
Boc-L-β-homoaspartic acid-γ-benzyl ester
Boc-L-beta-homoaspartic acid-gamma-benzyl ester

Hazard: Irritant

Formula: C₁₇H₂₃NO₆
MW: 337.38 g/mol
MDL Number: MFCD01862861
CAS Number: 254101-10-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR