3-(Boc-amino)-3-(4-methoxyphenyl)propionic acid

Supplier: Matrix Scientific

042275-250MG 042275-1G
101855-162EA 210.77 USD
101855-162 101855-164
3-(Boc-amino)-3-(4-methoxyphenyl)propionic acid
3-(Boc-amino)-3-(4-methoxyphenyl)propionic acid

Hazard: Irritant

Formula: C₁₅H₂₁NO₄
MDL Number: MFCD02090694
CAS Number: 96363-20-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR