Boc-3-amino-3-(3-bromophenyl)propionic acid

Supplier: Matrix Scientific

042273-1G 042273-5G
101855-154EA 504.62 USD
101855-154 101855-156
Boc-3-amino-3-(3-bromophenyl)propionic acid
Boc-3-amino-3-(3-bromophenyl)propionic acid

Hazard code(s): Irritant

Formula: C₁₄H₁₈BrNO₄
MDL Number: MFCD02090700
CAS Number: 284493-58-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR