3-(Boc-amino)-3-(2-chlorophenyl)propionic acid

Supplier: Matrix Scientific

Synonyms: 3-((tert-Butoxycarbonyl)amino)-3-(2-chlorophenyl)propanoic acid

042271-1G 042271-5G
101855-150EA 661.54 USD
101855-150 101855-152
3-(Boc-amino)-3-(2-chlorophenyl)propionic acid
3-(Boc-amino)-3-(2-chlorophenyl)propionic acid

Hazard: Irritant

Formula: C₁₄H₁₈ClNO₄
MW: 299.7500 g/mol
MDL Number: MFCD02090709
CAS Number: 284493-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR