Boc-isoguvacine

Supplier: Matrix Scientific

Synonyms: 1-Boc-1,2,3,6-tetrahydropyridine-4-carboxylic Acid

042270-100MG 042270-500MG
101855-146EA 547.69 USD
101855-146 101855-148
Boc-isoguvacine
Boc-isoguvacine

Hazard: Irritant

Formula: C₁₁H₁₇NO₄
MW: 227.26 g/mol
MDL Number: MFCD02682417
CAS Number: 70684-84-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR