Boc-(S)-3-amino-5-phenylpentanoic acid
Supplier: Matrix Scientific
042267-250MG
042267-1G
101855-134EA
275.38
USD
101855-134
101855-136
Boc-(S)-3-amino-5-phenylpentanoic acid
Boc-(S)-3-amino-5-phenylpentanoic acid
Hazard: Irritant
Formula:
C₁₆H₂₃NO₄ MW: 293.37 g/mol |
MDL Number:
MFCD01076264 CAS Number: 218608-84-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...