Boc-(S)-3-amino-4-(4-trifluoromethylphenyl)butyric acid
Supplier: Matrix Scientific
Synonyms:
(S)-3-((tert-Butoxycarbonyl)amino)-4-(4-(trifluoromethyl)phenyl)butanoic acid
042263-250MG
042263-1G
101855-118EA
412.31
USD
101855-118
101855-120
Boc-(S)-3-amino-4-(4-trifluoromethylphenyl)butyric acid
Boc-(S)-3-amino-4-(4-trifluoromethylphenyl)butyric acid
Hazard: Irritant
Formula:
C₁₆H₂₀F₃NO₄ MW: 347.34 g/mol |
MDL Number:
MFCD01861070 CAS Number: 270065-80-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...